Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: adh2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001111940.1 | NP_001105410.1 | 893 | 60 | ||
GenBank mRNA | AY104300.2 | - | 888 | 60 | ||
BT041397.1 | - | 550 | 60 | |||
X01965.1 | - | 893 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Zea mays |