Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gpa1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001111944.1 | NP_001105414.1 | 162 | 90 | ||
GenBank mRNA | X07157.1 | - | 102 | 90 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Zea mays |