Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: hir1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001112153.1 | NP_001105623.1 | 593 | 89 | ||
GenBank mRNA | AF236373.1 | - | 593 | 89 | ||
AJ292360.1 | - | 573 | 89 | |||
BT055443.1 | - | 585 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Zea mays |