Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ccp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001112215.1 | NP_001105685.1 | 614 | 81 | ||
GenBank mRNA | AY104312.2 | - | 675 | 81 | ||
D45402.1 | - | 614 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Protease Recommended Assay Zea mays |