Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sps1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001112224.1 | NP_001105694.1 | 1678 | 65 | ||
GenBank mRNA | AY109574.1 | - | 1678 | 65 | ||
M97550.1 | - | 1678 | 65 |