Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sgo1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001112321.1 | NP_001105791.1 | 993 | 86 | ||
GenBank mRNA | AY964185.1 | - | 993 | 86 |