Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: bf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001137483.1 | NP_001130955.1 | 522 | 71 | ||
GenBank mRNA | BT034353.1 | - | 200 | 71 | ||
BT067678.1 | - | 477 | 71 | |||
EU963353.1 | - | 522 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Zea mays |