Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: bf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001137483.1 | NP_001130955.1 | 955 | 96 | ||
GenBank mRNA | BT034353.1 | - | 633 | 96 | ||
BT067678.1 | - | 910 | 96 | |||
EU963353.1 | - | 955 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Zea mays |