Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rph1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001138140.1 | NP_001131612.1 | 329 | 79 | ||
GenBank mRNA | BT035096.1 | - | 329 | 79 | ||
EU960828.1 | - | 385 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Zea mays |