Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAGE | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001139139.1 | NP_001132611.1 | 994 | 90 | ||
GenBank mRNA | BT036524.1 | - | 994 | 90 | ||
BT038139.1 | - | 782 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Recommended Assay Zea mays |