Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tua6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001112118.1 | NP_001105588.1 | 1453 | 71 | ||
GenBank mRNA | AJ420857.1 | - | 672 | 71 | ||
BT068904.1 | - | 1659 | 71 | |||
X63178.1 | - | 1453 | 71 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Zea mays |