Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: prf5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001112152.1 | NP_001105622.1 | 655 | 87 | ||
GenBank mRNA | AF201459.1 | - | 655 | 87 | ||
BT041006.1 | - | 651 | 87 | |||
EU958791.1 | - | 663 | 87 | |||
EU960195.1 | - | 662 | 87 | |||
EU967239.1 | - | 731 | 87 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Zea mays |