Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cts1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001152570.1 | NP_001146042.1 | 2050 | 88 | ||
GenBank mRNA | BT054477.1 | - | 2050 | 88 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Zea mays |