Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCCTCTACGGCGCAGCTCGTCAAGC[C/T]GAAGGCTTTTTTCAAGGCATTCTTA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
OR6B3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
OR6B3 - olfactory receptor family 6 subfamily B member 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_173351.1 | 1313 | Missense Mutation | AGC,GGC | S,G 306 | NP_775486.1 | |
XM_011510671.2 | 1313 | Missense Mutation | AGC,GGC | S,G 413 | XP_011508973.1 |
PRR21 - proline rich 21 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |