Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGTAATCTGAGGCATGTCACTCGTA[C/T]TCGCATTCAAGGGCAGTCCTGGGTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611338 MIM: 188345 MIM: 608525 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ATG4B PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ATG4B - autophagy related 4B cysteine peptidase | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
DTYMK - deoxythymidylate kinase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001165031.1 | Intron | NP_001158503.1 | ||||
NM_001320902.1 | Intron | NP_001307831.1 | ||||
NM_001320903.1 | Intron | NP_001307832.1 | ||||
NM_001320904.1 | Intron | NP_001307833.1 | ||||
NM_001320905.1 | Intron | NP_001307834.1 | ||||
NM_012145.3 | Intron | NP_036277.2 |
ING5 - inhibitor of growth family member 5 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |