Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AATTGTTTTTTGTTAAATTAAAGCC[G/T]TAAGTAGTCTTAAATGATGTAGCAA
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 194553 | |||||||||||||||||||||||
Literature Links: |
ZNF80 PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
ZNF80 - zinc finger protein 80 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_007136.3 | 1846 | UTR 3 | NP_009067.2 | |||
XM_017007132.1 | 1846 | UTR 3 | XP_016862621.1 | |||
XM_017007133.1 | 1846 | UTR 3 | XP_016862622.1 | |||
XM_017007134.1 | 1846 | UTR 3 | XP_016862623.1 | |||
XM_017007135.1 | 1846 | Intron | XP_016862624.1 | |||
XM_017007136.1 | 1846 | Intron | XP_016862625.1 |