Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAATGGGTTCCTTTTATTTCTGCAA[C/T]AACATTGGGATTTTAATAGGGATTG
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
||||||||||||||||||||||||
Literature Links: |
CCDC14 PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
CCDC14 - coiled-coil domain containing 14 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001308317.1 | Intron | NP_001295246.1 | ||||
NM_022757.4 | Intron | NP_073594.4 | ||||
XM_005247710.4 | Intron | XP_005247767.1 | ||||
XM_005247711.4 | Intron | XP_005247768.1 | ||||
XM_005247713.2 | Intron | XP_005247770.1 | ||||
XM_005247714.2 | Intron | XP_005247771.1 | ||||
XM_005247715.4 | Intron | XP_005247772.1 | ||||
XM_005247716.4 | Intron | XP_005247773.1 | ||||
XM_006713731.3 | Intron | XP_006713794.1 | ||||
XM_006713733.3 | Intron | XP_006713796.1 | ||||
XM_011513081.2 | Intron | XP_011511383.1 | ||||
XM_017007056.1 | Intron | XP_016862545.1 | ||||
XM_017007057.1 | Intron | XP_016862546.1 | ||||
XM_017007058.1 | Intron | XP_016862547.1 | ||||
XM_017007059.1 | Intron | XP_016862548.1 | ||||
XM_017007060.1 | Intron | XP_016862549.1 | ||||
XM_017007061.1 | Intron | XP_016862550.1 |