Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTGGAGTTAGATGATTGCCACAGAC[A/C]TGGTAAATTCCTCAAACTCCTTAAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
EFCAB12 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
EFCAB12 - EF-hand calcium binding domain 12 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_207307.2 | 1623 | Missense Mutation | AGG,ATG | R,M 487 | NP_997190.1 | |
XM_011513293.2 | 1623 | Missense Mutation | AGG,ATG | R,M 525 | XP_011511595.1 | |
XM_011513294.2 | 1623 | Missense Mutation | AGG,ATG | R,M 524 | XP_011511596.1 | |
XM_011513295.2 | 1623 | Missense Mutation | AGG,ATG | R,M 510 | XP_011511597.1 | |
XM_011513297.2 | 1623 | Nonsense Mutation | GGA,TGA | G,* 474 | XP_011511599.1 | |
XM_011513298.1 | 1623 | Missense Mutation | AGG,ATG | R,M 240 | XP_011511600.1 | |
XM_011513299.2 | 1623 | Missense Mutation | AGG,ATG | R,M 233 | XP_011511601.1 |
RPL32P3 - ribosomal protein L32 pseudogene 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SNORA7B - small nucleolar RNA, H/ACA box 7B | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |