Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAACATGTTTGCAGTTTCTCCAAAA[C/T]AGACTCATCTTTCATCAGGTCTTTG
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 603212 | |||||||||||||||||||||||
Literature Links: |
BFSP2 PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
BFSP2 - beaded filament structural protein 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003571.3 | 100 | Intron | NP_003562.1 | |||
XM_017007315.1 | 100 | UTR 5 | XP_016862804.1 |
TMEM108 - transmembrane protein 108 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001136469.2 | 100 | Intron | NP_001129941.1 | |||
NM_001282865.1 | 100 | Intron | NP_001269794.1 | |||
NM_023943.3 | 100 | Intron | NP_076432.1 | |||
XM_011513097.2 | 100 | Intron | XP_011511399.1 | |||
XM_011513098.2 | 100 | Intron | XP_011511400.1 | |||
XM_017007080.1 | 100 | Intron | XP_016862569.1 | |||
XM_017007081.1 | 100 | Intron | XP_016862570.1 | |||
XM_017007082.1 | 100 | Intron | XP_016862571.1 |