Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACATTCAGTAAGTTATACTGGAGGC[C/T]CAAATTTCATGCTCCATGTAATTAT
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 607994 | |||||||||||||||||||||||
Literature Links: |
XRN1 PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
XRN1 - 5'-3' exoribonuclease 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001282857.1 | 5300 | UTR 3 | NP_001269786.1 | |||
NM_001282859.1 | 5300 | Intron | NP_001269788.1 | |||
NM_019001.4 | 5300 | UTR 3 | NP_061874.3 | |||
XM_006713673.2 | 5300 | Intron | XP_006713736.1 | |||
XM_011512919.2 | 5300 | UTR 3 | XP_011511221.1 | |||
XM_011512920.2 | 5300 | UTR 3 | XP_011511222.1 | |||
XM_011512922.2 | 5300 | UTR 3 | XP_011511224.1 | |||
XM_017006640.1 | 5300 | UTR 3 | XP_016862129.1 | |||
XM_017006641.1 | 5300 | UTR 3 | XP_016862130.1 | |||
XM_017006642.1 | 5300 | Intron | XP_016862131.1 |