Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGCATGGCTTCTGTAAAGGCACGTT[A/G]CTTTTTATTATAGTAATATTGTGGG
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 602734 | |||||||||||||||||||||||
Literature Links: |
PLS1 PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
PLS1 - plastin 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001145319.1 | Intron | NP_001138791.1 | ||||
NM_001172312.1 | Intron | NP_001165783.1 | ||||
NM_002670.2 | Intron | NP_002661.2 | ||||
XM_006713660.3 | Intron | XP_006713723.1 | ||||
XM_011512900.2 | Intron | XP_011511202.1 | ||||
XM_011512901.1 | Intron | XP_011511203.1 | ||||
XM_011512903.2 | Intron | XP_011511205.1 | ||||
XM_017006626.1 | Intron | XP_016862115.1 | ||||
XM_017006627.1 | Intron | XP_016862116.1 |