Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AACTTCTACATTATAGGTTGTTTTT[G/T]CCCTGGGTGGAATATTTAATGCTTT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
MFSD1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
MFSD1 - major facilitator superfamily domain containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001167903.1 | 578 | Missense Mutation | GCC,TCC | A,S 138 | NP_001161375.1 | |
NM_001289406.1 | 578 | Missense Mutation | GCC,TCC | A,S 31 | NP_001276335.1 | |
NM_001289407.1 | 578 | Missense Mutation | GCC,TCC | A,S 55 | NP_001276336.1 | |
NM_022736.2 | 578 | Missense Mutation | GCC,TCC | A,S 177 | NP_073573.2 | |
XM_006713730.2 | 578 | Missense Mutation | GCC,TCC | A,S 138 | XP_006713793.1 |