Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGTCTGTAACACGTAAATAGGATAG[A/G]TAGAATACTTTTTCAACTTCTGAAC
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 606042 | |||||||||||||||||||||||
Literature Links: |
LRRC34 PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
LRRC34 - leucine rich repeat containing 34 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001172779.1 | 1784 | Intron | NP_001166250.1 | |||
NM_001172780.1 | 1784 | Intron | NP_001166251.1 | |||
NM_153353.4 | 1784 | Intron | NP_699184.2 | |||
XM_006713508.3 | 1784 | UTR 3 | XP_006713571.1 | |||
XM_011512442.2 | 1784 | Intron | XP_011510744.1 | |||
XM_017005746.1 | 1784 | Intron | XP_016861235.1 |
MYNN - myoneurin | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |