Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAGTTTCTCCTGTCACTACCACAGT[C/T]TCAAGTTCTTGAGGACCACTGCTTT
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
||||||||||||||||||||||||
Literature Links: |
ZBTB11 PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
ZBTB11 - zinc finger and BTB domain containing 11 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014415.3 | 4567 | Silent Mutation | GAA,GAG | E,E 984 | NP_055230.2 | |
XM_011512689.2 | 4567 | Silent Mutation | GAA,GAG | E,E 919 | XP_011510991.1 | |
XM_011512690.2 | 4567 | Silent Mutation | GAA,GAG | E,E 700 | XP_011510992.1 |