Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TATCAAGAATTCAATTTTAAAGCAA[A/G]GAGAAAACTACATAACAGAAACTTG
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 608806 | |||||||||||||||||||||||
Literature Links: |
NAALADL2 PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
NAALADL2 - N-acetylated alpha-linked acidic dipeptidase-like 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_207015.2 | Intron | NP_996898.2 | ||||
XM_006713560.2 | Intron | XP_006713623.1 | ||||
XM_011512612.2 | Intron | XP_011510914.1 | ||||
XM_011512613.1 | Intron | XP_011510915.1 | ||||
XM_011512615.2 | Intron | XP_011510917.1 | ||||
XM_011512616.2 | Intron | XP_011510918.1 | ||||
XM_011512617.2 | Intron | XP_011510919.1 | ||||
XM_017006070.1 | Intron | XP_016861559.1 | ||||
XM_017006071.1 | Intron | XP_016861560.1 | ||||
XM_017006072.1 | Intron | XP_016861561.1 | ||||
XM_017006073.1 | Intron | XP_016861562.1 | ||||
XM_017006074.1 | Intron | XP_016861563.1 | ||||
XM_017006075.1 | Intron | XP_016861564.1 | ||||
XM_017006076.1 | Intron | XP_016861565.1 | ||||
XM_017006077.1 | Intron | XP_016861566.1 | ||||
XM_017006078.1 | Intron | XP_016861567.1 | ||||
XM_017006079.1 | Intron | XP_016861568.1 | ||||
XM_017006080.1 | Intron | XP_016861569.1 | ||||
XM_017006081.1 | Intron | XP_016861570.1 | ||||
XM_017006082.1 | Intron | XP_016861571.1 | ||||
XM_017006083.1 | Intron | XP_016861572.1 |