Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCAGTGTGTGGAAATACCAGACGTA[G/T]AACTGGTAGTGGAGGGAGCGGCTGA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 608750 MIM: 610145 MIM: 611620 | ||||||||||||||||||||
Literature Links: |
ALG3 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ALG3 - ALG3, alpha-1,3- mannosyltransferase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001006941.2 | 996 | Missense Mutation | TTA,TTC | L,F 312 | NP_001006942.1 | |
NM_005787.5 | 996 | Missense Mutation | TTA,TTC | L,F 360 | NP_005778.1 | |
XM_011512322.1 | 996 | Missense Mutation | TTA,TTC | L,F 327 | XP_011510624.1 | |
XM_011512323.2 | 996 | Missense Mutation | TTA,TTC | L,F 320 | XP_011510625.1 |
ECE2 - endothelin converting enzyme 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIR1224 - microRNA 1224 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
VWA5B2 - von Willebrand factor A domain containing 5B2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001320373.1 | 996 | Intron | NP_001307302.1 | |||
NM_138345.2 | 996 | Intron | NP_612354.1 | |||
XM_011513288.2 | 996 | Intron | XP_011511590.1 | |||
XM_011513289.2 | 996 | Intron | XP_011511591.1 | |||
XM_011513290.2 | 996 | Intron | XP_011511592.1 | |||
XM_011513291.2 | 996 | Intron | XP_011511593.1 | |||
XM_011513292.2 | 996 | Intron | XP_011511594.1 |