Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTTACTTATTTTTTTAAATTAATAT[C/T]CACAATCAGTGATTCAAAAAGCAGC
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 600521 | |||||||||||||||||||||||
Literature Links: |
LOC101929106 PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
LOC101929106 - uncharacterized LOC101929106 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MASP1 - mannan binding lectin serine peptidase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001031849.2 | 5036 | Intron | NP_001027019.1 | |||
NM_001879.5 | 5036 | UTR 3 | NP_001870.3 | |||
NM_139125.3 | 5036 | Intron | NP_624302.1 | |||
XM_006713701.2 | 5036 | Intron | XP_006713764.1 | |||
XM_011512989.2 | 5036 | Intron | XP_011511291.1 | |||
XM_011512990.2 | 5036 | Intron | XP_011511292.1 | |||
XM_011512991.2 | 5036 | Intron | XP_011511293.1 | |||
XM_017006869.1 | 5036 | Intron | XP_016862358.1 | |||
XM_017006870.1 | 5036 | Intron | XP_016862359.1 | |||
XM_017006871.1 | 5036 | Intron | XP_016862360.1 | |||
XM_017006872.1 | 5036 | Intron | XP_016862361.1 |