Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGAGGTGTAGGTGGTCACCTCCTC[C/T]TCCAGCAGCTTCTCGCTGGGCTTCC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 609138 | ||||||||||||||||||||
Literature Links: |
LOC100131635 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
LOC100131635 - hCG1645011-like | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RTP2 - receptor transporter protein 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001004312.2 | 1093 | Silent Mutation | GAA,GAG | E,E 173 | NP_001004312.2 | |
XM_017006301.1 | 1093 | Silent Mutation | GAA,GAG | E,E 173 | XP_016861790.1 | |
XM_017006302.1 | 1093 | Silent Mutation | GAA,GAG | E,E 173 | XP_016861791.1 |