Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTGAGTTTCATTCAACCAATGTCCA[C/T]TGAGCCCCTTCTGTGGGTCAGCCCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611195 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
JAKMIP1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
JAKMIP1 - janus kinase and microtubule interacting protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001099433.1 | Intron | NP_001092903.1 | ||||
NM_001306133.1 | Intron | NP_001293062.1 | ||||
NM_001306134.1 | Intron | NP_001293063.1 | ||||
NM_144720.3 | Intron | NP_653321.1 | ||||
XM_011513400.2 | Intron | XP_011511702.1 | ||||
XM_017007788.1 | Intron | XP_016863277.1 | ||||
XM_017007789.1 | Intron | XP_016863278.1 | ||||
XM_017007790.1 | Intron | XP_016863279.1 | ||||
XM_017007791.1 | Intron | XP_016863280.1 | ||||
XM_017007792.1 | Intron | XP_016863281.1 | ||||
XM_017007793.1 | Intron | XP_016863282.1 | ||||
XM_017007794.1 | Intron | XP_016863283.1 | ||||
XM_017007795.1 | Intron | XP_016863284.1 | ||||
XM_017007796.1 | Intron | XP_016863285.1 | ||||
XM_017007797.1 | Intron | XP_016863286.1 | ||||
XM_017007798.1 | Intron | XP_016863287.1 | ||||
XM_017007799.1 | Intron | XP_016863288.1 |