Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACAGTCCTGGTCGTCGCCGCTGCCC[C/G]GCCGGGGTGCGAGTTTGAGCGGAAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603105 MIM: 606921 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CPZ PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CPZ - carboxypeptidase Z | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001014447.2 | 230 | Missense Mutation | CCG,CGG | P,R 19 | NP_001014447.1 | |
NM_001014448.2 | 230 | UTR 5 | NP_001014448.1 | |||
NM_003652.3 | 230 | Missense Mutation | CCG,CGG | P,R 19 | NP_003643.2 |
GPR78 - G protein-coupled receptor 78 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |