Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTCATTTTGTGGCACTTTGTTACAG[C/T]ACCCCTAGAAAACTTATACACCGAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606619 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GBA3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GBA3 - glucosylceramidase beta 3 (gene/pseudogene) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001128432.2 | Intron | NP_001121904.1 | ||||
NM_001277225.1 | Intron | NP_001264154.1 | ||||
NM_020973.4 | Intron | NP_066024.1 |