Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGAATACTGACATACAAAATAAAGA[C/T]AGTGCAAGAAGAACTGCAATCTATT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611799 MIM: 606280 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LCORL PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LCORL - ligand dependent nuclear receptor corepressor like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001166139.2 | 5833 | Intron | NP_001159611.1 | |||
NM_153686.8 | 5833 | UTR 3 | NP_710153.2 | |||
XM_011513821.2 | 5833 | UTR 3 | XP_011512123.1 | |||
XM_011513822.2 | 5833 | UTR 3 | XP_011512124.1 | |||
XM_011513823.2 | 5833 | UTR 3 | XP_011512125.1 | |||
XM_011513825.2 | 5833 | Intron | XP_011512127.1 | |||
XM_011513826.2 | 5833 | UTR 3 | XP_011512128.1 | |||
XM_017007961.1 | 5833 | UTR 3 | XP_016863450.1 | |||
XM_017007962.1 | 5833 | UTR 3 | XP_016863451.1 | |||
XM_017007963.1 | 5833 | UTR 3 | XP_016863452.1 | |||
XM_017007964.1 | 5833 | Intron | XP_016863453.1 | |||
XM_017007965.1 | 5833 | UTR 3 | XP_016863454.1 |
NCAPG - non-SMC condensin I complex subunit G | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_022346.4 | 5833 | Intron | NP_071741.2 | |||
XM_011513876.2 | 5833 | Intron | XP_011512178.1 | |||
XM_017008543.1 | 5833 | Intron | XP_016864032.1 | |||
XM_017008544.1 | 5833 | Intron | XP_016864033.1 |