Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGTGATGATTATTGTGCTTGAAGG[A/T]TGGGTTTCTAATTCTGTCATAAAAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610472 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LPCAT1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LPCAT1 - lysophosphatidylcholine acyltransferase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
XM_005248373.3 | 2388 | UTR 3 | XP_005248430.1 | |||
XM_011514132.1 | 2388 | UTR 3 | XP_011512434.1 | |||
XM_011514134.1 | 2388 | UTR 3 | XP_011512436.1 | |||
XM_017009879.1 | 2388 | UTR 3 | XP_016865368.1 |