Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTCAATGAGAGACAATATTCATAAT[C/T]AAAACAGTGGTAATAGCATAAATGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604555 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CDH10 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CDH10 - cadherin 10 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001317222.1 | Intron | NP_001304151.1 | ||||
NM_001317224.1 | Intron | NP_001304153.1 | ||||
NM_006727.4 | Intron | NP_006718.2 | ||||
XM_011513923.2 | Intron | XP_011512225.1 | ||||
XM_017008915.1 | Intron | XP_016864404.1 |