Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAGCCATATTGCAACTACATATAAA[A/G]GTAGTTATAGATTCTTTTGTATTTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606517 MIM: 601057 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
AHRR PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
AHRR - aryl-hydrocarbon receptor repressor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001242412.1 | Intron | NP_001229341.1 | ||||
NM_020731.4 | Intron | NP_065782.2 |
PDCD6 - programmed cell death 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001267556.1 | Intron | NP_001254485.1 | ||||
NM_001267557.1 | Intron | NP_001254486.1 | ||||
NM_001267558.1 | Intron | NP_001254487.1 | ||||
NM_001267559.1 | Intron | NP_001254488.1 | ||||
NM_013232.3 | Intron | NP_037364.1 |