Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCCGCGGGAGGGCGCGCTGGTCTCC[C/G]GCGTTACCCCAAGCTCCCAGTGTGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608828 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
C5orf22 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
C5orf22 - chromosome 5 open reading frame 22 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_018356.2 | 342 | Missense Mutation | CCG,CGG | P,R 12 | NP_060826.2 | |
XM_006714479.2 | 342 | UTR 5 | XP_006714542.1 | |||
XM_006714480.3 | 342 | UTR 5 | XP_006714543.1 | |||
XM_011514062.2 | 342 | Missense Mutation | CCG,CGG | P,R 12 | XP_011512364.1 | |
XM_017009607.1 | 342 | Missense Mutation | CCG,CGG | P,R 12 | XP_016865096.1 | |
XM_017009608.1 | 342 | Missense Mutation | CCG,CGG | P,R 12 | XP_016865097.1 | |
XM_017009609.1 | 342 | UTR 5 | XP_016865098.1 | |||
XM_017009610.1 | 342 | UTR 5 | XP_016865099.1 | |||
XM_017009611.1 | 342 | UTR 5 | XP_016865100.1 | |||
XM_017009612.1 | 342 | UTR 5 | XP_016865101.1 | |||
XM_017009613.1 | 342 | UTR 5 | XP_016865102.1 | |||
XM_017009614.1 | 342 | UTR 5 | XP_016865103.1 | |||
XM_017009615.1 | 342 | UTR 5 | XP_016865104.1 | |||
XM_017009616.1 | 342 | UTR 5 | XP_016865105.1 |
DROSHA - drosha ribonuclease III | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001100412.1 | 342 | Intron | NP_001093882.1 | |||
NM_013235.4 | 342 | Intron | NP_037367.3 | |||
XM_005248291.3 | 342 | Intron | XP_005248348.1 | |||
XM_005248292.3 | 342 | Intron | XP_005248349.1 | |||
XM_005248293.3 | 342 | Intron | XP_005248350.1 | |||
XM_005248294.3 | 342 | Intron | XP_005248351.1 | |||
XM_011514033.2 | 342 | Intron | XP_011512335.1 | |||
XM_017009399.1 | 342 | Intron | XP_016864888.1 | |||
XM_017009400.1 | 342 | Intron | XP_016864889.1 | |||
XM_017009401.1 | 342 | Intron | XP_016864890.1 | |||
XM_017009402.1 | 342 | Intron | XP_016864891.1 |