Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGATACCTCTTTTGTTAATGGCTAC[A/G]TGTTATTCTTTTTGTGTGTGTGTGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601743 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
OSMR PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
OSMR - oncostatin M receptor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001168355.2 | Intron | NP_001161827.1 | ||||
NM_001323504.1 | Intron | NP_001310433.1 | ||||
NM_001323505.1 | Intron | NP_001310434.1 | ||||
NM_001323506.1 | Intron | NP_001310435.1 | ||||
NM_001323507.1 | Intron | NP_001310436.1 | ||||
NM_003999.2 | Intron | NP_003990.1 | ||||
XM_005248384.1 | Intron | XP_005248441.1 | ||||
XM_005248386.2 | Intron | XP_005248443.1 | ||||
XM_005248387.2 | Intron | XP_005248444.1 | ||||
XM_011514161.2 | Intron | XP_011512463.1 | ||||
XM_017010019.1 | Intron | XP_016865508.1 | ||||
XM_017010020.1 | Intron | XP_016865509.1 |
OSMR-AS1 - OSMR antisense RNA 1 (head to head) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |