Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTGGTCAAATTGAGAACAACGGCA[C/T]CTAAAGTTAATAGGAGACCATGTTA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
MROH2B PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MROH2B - maestro heat like repeat family member 2B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_173489.4 | 5062 | Missense Mutation | GAT,GGT | D,G 1529 | NP_775760.3 | |
XM_011513952.1 | 5062 | Missense Mutation | GAT,GGT | D,G 1529 | XP_011512254.1 | |
XM_011513953.1 | 5062 | Missense Mutation | GAT,GGT | D,G 1467 | XP_011512255.1 |