Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTAAGTGCCTGCTACATAGGCACT[C/G]TATGTCCACATGGAAACCATAGCAT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 617016 | ||||||||||||||||||||
Literature Links: |
PLCXD3 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
PLCXD3 - phosphatidylinositol specific phospholipase C X domain containing 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001005473.2 | 3866 | UTR 3 | NP_001005473.1 | |||
XM_017009436.1 | 3866 | Intron | XP_016864925.1 | |||
XM_017009437.1 | 3866 | Intron | XP_016864926.1 | |||
XM_017009438.1 | 3866 | Intron | XP_016864927.1 |