Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGACGAGGCCAACATAAAGACAGCA[A/G]GCATCAGCAGAGGATGGAGACAGCG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606517 MIM: 608186 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
AHRR PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
AHRR - aryl-hydrocarbon receptor repressor | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
EXOC3 - exocyst complex component 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_007277.4 | Intron | NP_009208.2 | ||||
XM_011513948.2 | Intron | XP_011512250.1 | ||||
XM_011513949.2 | Intron | XP_011512251.1 | ||||
XM_011513950.2 | Intron | XP_011512252.1 | ||||
XM_011513951.2 | Intron | XP_011512253.1 | ||||
XM_017009000.1 | Intron | XP_016864489.1 | ||||
XM_017009001.1 | Intron | XP_016864490.1 | ||||
XM_017009002.1 | Intron | XP_016864491.1 | ||||
XM_017009003.1 | Intron | XP_016864492.1 | ||||
XM_017009004.1 | Intron | XP_016864493.1 | ||||
XM_017009005.1 | Intron | XP_016864494.1 | ||||
XM_017009006.1 | Intron | XP_016864495.1 |
EXOC3-AS1 - EXOC3 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |