Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAATTTGCTGTGGTACTTACGGTAC[C/T]ATTTGCTTCCAAAATTCATGTGATT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603390 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PDE8B PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PDE8B - phosphodiesterase 8B | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
WDR41 - WD repeat domain 41 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_018268.3 | 3041 | UTR 3 | NP_060738.2 | |||
XM_005248551.4 | 3041 | UTR 3 | XP_005248608.1 | |||
XM_005248552.3 | 3041 | Intron | XP_005248609.1 | |||
XM_011543505.2 | 3041 | UTR 3 | XP_011541807.1 | |||
XM_011543507.2 | 3041 | Intron | XP_011541809.1 | |||
XM_017009606.1 | 3041 | UTR 3 | XP_016865095.1 |