Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTACCATCCTTTCTTCAGTGTTTAT[A/G]CTCAGTTCCTGAAATGCTTTTGAGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610800 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ATG10 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ATG10 - autophagy related 10 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001131028.1 | Intron | NP_001124500.1 | ||||
NM_031482.4 | Intron | NP_113670.1 | ||||
XM_005248610.4 | Intron | XP_005248667.1 | ||||
XM_005248611.4 | Intron | XP_005248668.1 | ||||
XM_005248612.3 | Intron | XP_005248669.1 | ||||
XM_011543660.2 | Intron | XP_011541962.1 | ||||
XM_011543661.2 | Intron | XP_011541963.1 | ||||
XM_017009944.1 | Intron | XP_016865433.1 |