Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAATAATAAGAATTTTGAAAGTTGC[A/T]TCTTTGTTTCTGCATTAGGGATTCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 115435 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
HAPLN1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
HAPLN1 - hyaluronan and proteoglycan link protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001884.3 | 2321 | UTR 3 | NP_001875.1 | |||
XM_011543168.2 | 2321 | UTR 3 | XP_011541470.1 | |||
XM_017009051.1 | 2321 | UTR 3 | XP_016864540.1 | |||
XM_017009052.1 | 2321 | UTR 3 | XP_016864541.1 | |||
XM_017009053.1 | 2321 | UTR 3 | XP_016864542.1 | |||
XM_017009054.1 | 2321 | UTR 3 | XP_016864543.1 |