Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTCTATAAACTCTGGTATATATGT[G/T]AAGGAGTTACTGTTGGCTGGCTTGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600694 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
IL6ST PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
IL6ST - interleukin 6 signal transducer | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001190981.1 | 8114 | UTR 3 | NP_001177910.1 | |||
NM_002184.3 | 8114 | UTR 3 | NP_002175.2 | |||
NM_175767.2 | 8114 | UTR 3 | NP_786943.1 | |||
XM_017009441.1 | 8114 | Intron | XP_016864930.1 | |||
XM_017009442.1 | 8114 | Intron | XP_016864931.1 |