Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAGATCCCTGATTTTGTGTCTGTTT[G/T]GGGAAAGATGCTGAAAATTTTGTCC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 615189 | ||||||||||||||||||||
Literature Links: |
ANKRD55 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ANKRD55 - ankyrin repeat domain 55 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_024669.2 | 1121 | Missense Mutation | CAA,CCA | Q,P 552 | NP_078945.2 | |
XM_011543646.1 | 1121 | Missense Mutation | CAA,CCA | Q,P 325 | XP_011541948.1 | |
XM_017009852.1 | 1121 | Missense Mutation | CAA,CCA | Q,P 561 | XP_016865341.1 | |
XM_017009853.1 | 1121 | Missense Mutation | CAA,CCA | Q,P 552 | XP_016865342.1 | |
XM_017009854.1 | 1121 | Missense Mutation | CAA,CCA | Q,P 399 | XP_016865343.1 |