Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAAGTCTATCTATCACTGATATCAG[C/T]GCCTTGGACCTGTGGAATTCTGAAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
OGFRL1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
OGFRL1 - opioid growth factor receptor like 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001324266.1 | 166 | Intron | NP_001311195.1 | |||
NM_024576.4 | 166 | Intron | NP_078852.3 | |||
XM_011536117.2 | 166 | Intron | XP_011534419.1 | |||
XM_011536118.2 | 166 | Intron | XP_011534420.1 | |||
XM_017011303.1 | 166 | Intron | XP_016866792.1 | |||
XM_017011304.1 | 166 | Intron | XP_016866793.1 | |||
XM_017011305.1 | 166 | Intron | XP_016866794.1 | |||
XM_017011306.1 | 166 | UTR 5 | XP_016866795.1 |