Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTTATAGGAAAGATGGTAAATACTC[A/G]TTTCTTTGCTTGAGTTGGAATTAAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615375 MIM: 612870 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
IRAK1BP1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
IRAK1BP1 - interleukin 1 receptor associated kinase 1 binding protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001010844.3 | 8052 | Intron | NP_001010844.1 | |||
XM_006715336.3 | 8052 | Intron | XP_006715399.1 | |||
XM_011535447.1 | 8052 | Intron | XP_011533749.1 | |||
XM_011535448.2 | 8052 | Intron | XP_011533750.1 | |||
XM_017010264.1 | 8052 | Intron | XP_016865753.1 | |||
XM_017010265.1 | 8052 | Intron | XP_016865754.1 |
PHIP - pleckstrin homology domain interacting protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_017934.5 | 8052 | UTR 3 | NP_060404.3 | |||
XM_005248729.4 | 8052 | Intron | XP_005248786.1 | |||
XM_011535918.2 | 8052 | Intron | XP_011534220.1 | |||
XM_011535919.1 | 8052 | Intron | XP_011534221.1 | |||
XM_017010989.1 | 8052 | Intron | XP_016866478.1 | |||
XM_017010990.1 | 8052 | Intron | XP_016866479.1 |