Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGTCGTCGCTACTCTGGTGGAACTC[A/G]GAGTTGGTTCTGGAGGCGGCGGACG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 190920 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
TPBG PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
TPBG - trophoblast glycoprotein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001166392.1 | 567 | Intron | NP_001159864.1 | |||
NM_006670.4 | 567 | UTR 5 | NP_006661.1 | |||
XM_011536097.2 | 567 | UTR 5 | XP_011534399.1 |