Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AATATTAGATACCTTAAATTGTTTA[C/T]AAGAAATCCTTGATGTTAAGATCCG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607923 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SNAP91 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SNAP91 - synaptosome associated protein 91 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001242792.1 | 2758 | Silent Mutation | TTA,TTG | L,L 907 | NP_001229721.1 | |
NM_001242793.1 | 2758 | Silent Mutation | TTA,TTG | L,L 877 | NP_001229722.1 | |
NM_001242794.1 | 2758 | Silent Mutation | TTA,TTG | L,L 600 | NP_001229723.1 | |
NM_001256717.1 | 2758 | Silent Mutation | TTA,TTG | L,L 816 | NP_001243646.1 | |
NM_001256718.1 | 2758 | Silent Mutation | TTA,TTG | L,L 783 | NP_001243647.1 | |
NM_014841.2 | 2758 | Silent Mutation | TTA,TTG | L,L 907 | NP_055656.1 | |
XM_005248770.4 | 2758 | Silent Mutation | TTA,TTG | L,L 907 | XP_005248827.1 | |
XM_006715615.1 | 2758 | Silent Mutation | TTA,TTG | L,L 907 | XP_006715678.1 | |
XM_011536265.1 | 2758 | Silent Mutation | TTA,TTG | L,L 907 | XP_011534567.1 | |
XM_011536266.1 | 2758 | Silent Mutation | TTA,TTG | L,L 907 | XP_011534568.1 | |
XM_011536269.1 | 2758 | Silent Mutation | TTA,TTG | L,L 902 | XP_011534571.1 | |
XM_011536271.1 | 2758 | Silent Mutation | TTA,TTG | L,L 893 | XP_011534573.1 | |
XM_011536273.2 | 2758 | Silent Mutation | TTA,TTG | L,L 881 | XP_011534575.1 | |
XM_011536275.2 | 2758 | Silent Mutation | TTA,TTG | L,L 879 | XP_011534577.1 | |
XM_011536276.2 | 2758 | Silent Mutation | TTA,TTG | L,L 871 | XP_011534578.1 | |
XM_011536277.1 | 2758 | Silent Mutation | TTA,TTG | L,L 849 | XP_011534579.1 | |
XM_017011554.1 | 2758 | Silent Mutation | TTA,TTG | L,L 905 | XP_016867043.1 | |
XM_017011555.1 | 2758 | Silent Mutation | TTA,TTG | L,L 905 | XP_016867044.1 | |
XM_017011556.1 | 2758 | Silent Mutation | TTA,TTG | L,L 905 | XP_016867045.1 | |
XM_017011557.1 | 2758 | Silent Mutation | TTA,TTG | L,L 902 | XP_016867046.1 | |
XM_017011558.1 | 2758 | Silent Mutation | TTA,TTG | L,L 902 | XP_016867047.1 | |
XM_017011559.1 | 2758 | Silent Mutation | TTA,TTG | L,L 902 | XP_016867048.1 | |
XM_017011560.1 | 2758 | Silent Mutation | TTA,TTG | L,L 902 | XP_016867049.1 | |
XM_017011561.1 | 2758 | Silent Mutation | TTA,TTG | L,L 902 | XP_016867050.1 | |
XM_017011562.1 | 2758 | Silent Mutation | TTA,TTG | L,L 902 | XP_016867051.1 | |
XM_017011563.1 | 2758 | Silent Mutation | TTA,TTG | L,L 900 | XP_016867052.1 | |
XM_017011564.1 | 2758 | Silent Mutation | TTA,TTG | L,L 900 | XP_016867053.1 | |
XM_017011565.1 | 2758 | Silent Mutation | TTA,TTG | L,L 900 | XP_016867054.1 | |
XM_017011566.1 | 2758 | Silent Mutation | TTA,TTG | L,L 893 | XP_016867055.1 | |
XM_017011567.1 | 2758 | Silent Mutation | TTA,TTG | L,L 893 | XP_016867056.1 | |
XM_017011568.1 | 2758 | Silent Mutation | TTA,TTG | L,L 891 | XP_016867057.1 | |
XM_017011569.1 | 2758 | Silent Mutation | TTA,TTG | L,L 888 | XP_016867058.1 | |
XM_017011570.1 | 2758 | Silent Mutation | TTA,TTG | L,L 881 | XP_016867059.1 | |
XM_017011571.1 | 2758 | Silent Mutation | TTA,TTG | L,L 881 | XP_016867060.1 | |
XM_017011572.1 | 2758 | Silent Mutation | TTA,TTG | L,L 881 | XP_016867061.1 | |
XM_017011573.1 | 2758 | Silent Mutation | TTA,TTG | L,L 881 | XP_016867062.1 | |
XM_017011574.1 | 2758 | Intron | XP_016867063.1 | |||
XM_017011575.1 | 2758 | Silent Mutation | TTA,TTG | L,L 879 | XP_016867064.1 | |
XM_017011576.1 | 2758 | Silent Mutation | TTA,TTG | L,L 879 | XP_016867065.1 | |
XM_017011577.1 | 2758 | Silent Mutation | TTA,TTG | L,L 879 | XP_016867066.1 | |
XM_017011578.1 | 2758 | Silent Mutation | TTA,TTG | L,L 879 | XP_016867067.1 | |
XM_017011579.1 | 2758 | Silent Mutation | TTA,TTG | L,L 874 | XP_016867068.1 | |
XM_017011580.1 | 2758 | Silent Mutation | TTA,TTG | L,L 871 | XP_016867069.1 | |
XM_017011581.1 | 2758 | Silent Mutation | TTA,TTG | L,L 871 | XP_016867070.1 | |
XM_017011582.1 | 2758 | Silent Mutation | TTA,TTG | L,L 871 | XP_016867071.1 | |
XM_017011583.1 | 2758 | Silent Mutation | TTA,TTG | L,L 866 | XP_016867072.1 | |
XM_017011584.1 | 2758 | Silent Mutation | TTA,TTG | L,L 866 | XP_016867073.1 | |
XM_017011585.1 | 2758 | Silent Mutation | TTA,TTG | L,L 866 | XP_016867074.1 | |
XM_017011586.1 | 2758 | Silent Mutation | TTA,TTG | L,L 866 | XP_016867075.1 | |
XM_017011587.1 | 2758 | Silent Mutation | TTA,TTG | L,L 866 | XP_016867076.1 | |
XM_017011588.1 | 2758 | Silent Mutation | TTA,TTG | L,L 866 | XP_016867077.1 | |
XM_017011589.1 | 2758 | Silent Mutation | TTA,TTG | L,L 857 | XP_016867078.1 | |
XM_017011590.1 | 2758 | Silent Mutation | TTA,TTG | L,L 843 | XP_016867079.1 |