Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAATGAGTATATTCATTGGGGACAA[C/T]GTTTTCGTCACTTGGGTTAAGAAAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 114610 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CNR1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CNR1 - cannabinoid receptor 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001160226.1 | 4325 | UTR 3 | NP_001153698.1 | |||
NM_001160258.1 | 4325 | UTR 3 | NP_001153730.1 | |||
NM_001160259.1 | 4325 | UTR 3 | NP_001153731.1 | |||
NM_016083.4 | 4325 | UTR 3 | NP_057167.2 | |||
NM_033181.3 | 4325 | UTR 3 | NP_149421.2 | |||
XM_006715330.3 | 4325 | UTR 3 | XP_006715393.1 | |||
XM_011535424.1 | 4325 | UTR 3 | XP_011533726.1 | |||
XM_011535425.1 | 4325 | UTR 3 | XP_011533727.1 | |||
XM_011535426.1 | 4325 | UTR 3 | XP_011533728.1 | |||
XM_011535428.1 | 4325 | UTR 3 | XP_011533730.1 | |||
XM_017010238.1 | 4325 | UTR 3 | XP_016865727.1 | |||
XM_017010239.1 | 4325 | UTR 3 | XP_016865728.1 | |||
XM_017010240.1 | 4325 | UTR 3 | XP_016865729.1 |